Merck
  • Microsomal cytochrome p450-mediated metabolism of protopanaxatriol ginsenosides: metabolite profile, reaction phenotyping, and structure-metabolism relationship.
Microsomal cytochrome p450-mediated metabolism of protopanaxatriol ginsenosides: metabolite profile, reaction phenotyping, and structure-metabolism relationship.
Drug metabolism and disposition: the biological fate of chemicals (2010-07-20)
Haiping Hao, Li Lai, Chaonan Zheng, Qiong Wang, Guo Yu, Xueyan Zhou, Liang Wu, Ping Gong, Guangji Wang
ZUSAMMENFASSUNG
Although the biotransformation of ginsenosides in the gastrointestinal tract has been extensively studied, much less is known about hepatic cytochrome P450 (P450)-catalyzed metabolism. The major aims of this study were to clarify the metabolic pathway and P450 isoforms involved and to explore the structure-metabolism relationship of protopanaxatriol (PPT)-type ginsenosides in hepatic microsomes. Efficient depletion of ginsenoside Rh1, Rg2, Rf, and PPT was found, whereas the elimination of Re and Rg1, characterized by a glucose substitution at the C20 hydroxy group, was negligible in microsomal incubation systems. Based on high-performance liquid chromatography hybrid ion trap and time-of-flight mass spectrometry analysis, the oxygenation metabolism on the C20 aliphatic branch chain was identified as the predominant metabolic pathway of PPT ginsenosides in both human and rat hepatic microsomes. By a comparison with authentic standards, the C24-25 double bond was identified as one of the oxygenation sites to produce the metabolites of C20-24 epoxide (ocotillol-type ginsenosides). Both chemical inhibition and human recombinant P450 isoform assays indicated that CYP3A4 was the predominant isozyme responsible for the oxygenation metabolism of PPT ginsenosides. Enzyme kinetic evaluations in rat and human hepatic microsomes and human recombinant CYP3A4 isozyme incubation systems showed generally consistent results in that the intrinsic clearance ranked as Rf ≤ Rg2 < Rh1 < PPT, closely correlating with logP values and the number of glycosyl substitutions. Results obtained from this study suggest that CYP3A4-catalyzed oxygenation metabolism plays an important role in the hepatic disposition of ginsenosides and that glycosyl substitution, especially at the C20 hydroxy group, determines their intrinsic clearances by CYP3A4.