Merck
  • Ultrafast deactivation processes in the 2-aminopyridine dimer and the adenine-thymine base pair: similarities and differences.
Ultrafast deactivation processes in the 2-aminopyridine dimer and the adenine-thymine base pair: similarities and differences.
The Journal of chemical physics (2010-08-17)
Yue-Jie Ai, Feng Zhang, Gang-Long Cui, Yi Luo, Wei-Hai Fang
ZUSAMMENFASSUNG
2-Aminopyridine dimer has frequently been used as a model system for studying photochemistry of DNA base pairs. We examine here the relevance of 2-aminopyridine dimer for a Watson-Crick adenine-thymine base pair by studying UV-light induced photodynamics along two main hydrogen bridges after the excitation to the localized (1)pi pi(*) excited-state. The respective two-dimensional potential-energy surfaces have been determined by time-dependent density functional theory with Coulomb-attenuated hybrid exchange-correlation functional (CAM-B3LYP). Different mechanistic aspects of the deactivation pathway have been analyzed and compared in detail for both systems, while the related reaction rates have also be obtained from Monte Carlo kinetic simulations. The limitations of the 2-aminopyridine dimer as a model system for the adenine-thymine base pair are discussed.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Aminopyridin, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
2-Aminopyridin, 99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
2-Aminopyridin, PESTANAL®, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
2-Aminopyridin, purum, ≥98.0% (NT)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.