Merck
  • Real-time two-dimensional imaging of potassium ion distribution using an ion semiconductor sensor with charged coupled device technology.
Real-time two-dimensional imaging of potassium ion distribution using an ion semiconductor sensor with charged coupled device technology.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2010-10-19)
Toshiaki Hattori, Yoshitomo Masaki, Kazuya Atsumi, Ryo Kato, Kazuaki Sawada
ZUSAMMENFASSUNG
Two-dimensional real-time observation of potassium ion distributions was achieved using an ion imaging device based on charge-coupled device (CCD) and metal-oxide semiconductor technologies, and an ion selective membrane. The CCD potassium ion image sensor was equipped with an array of 32 × 32 pixels (1024 pixels). It could record five frames per second with an area of 4.16 × 4.16 mm(2). Potassium ion images were produced instantly. The leaching of potassium ion from a 3.3 M KCl Ag/AgCl reference electrode was dynamically monitored in aqueous solution. The potassium ion selective membrane on the semiconductor consisted of plasticized poly(vinyl chloride) (PVC) with bis(benzo-15-crown-5). The addition of a polyhedral oligomeric silsesquioxane to the plasticized PVC membrane greatly improved adhesion of the membrane onto Si(3)N(4) of the semiconductor surface, and the potential response was stabilized. The potential response was linear from 10(-2) to 10(-5) M logarithmic concentration of potassium ion. The selectivity coefficients were K(K(+),Li(+))(pot) = 10(-2.85), K(K(+),Na(+))(pot) = 10(-2.30), K(K(+),Rb(+))(pot) =10(-1.16), and K(K(+),Cs(+))(pot) = 10(-2.05).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
15-Krone-5, 98%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.