Merck
Production of glutaconic acid in a recombinant Escherichia coli strain.
Applied and environmental microbiology (2010-11-03)
Ivana Djurdjevic, Oskar Zelder, Wolfgang Buckel
ZUSAMMENFASSUNG
The assembly of six genes that encode enzymes from glutamate-fermenting bacteria converted Escherichia coli into a glutaconate producer when grown anaerobically on a complex medium. The new anaerobic pathway starts with 2-oxoglutarate from general metabolism and proceeds via (R)-2-hydroxyglutarate, (R)-2-hydroxyglutaryl-coenzyme A (CoA), and (E)-glutaconyl-CoA to yield 2.7 ± 0.2 mM (E)-glutaconate in the medium.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glutakonsäure, 97% (T)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.