- [Analysis of essential oil from Mahonia duclouxiana].
[Analysis of essential oil from Mahonia duclouxiana].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2010-12-09)
Si-Xiang Liu, Bu-Ming Liu, Kai-Jia He, Xiao Lin, Wen-Jie Lu
PMID21137367
ZUSAMMENFASSUNG
To analyze the compositions of essential oil from Mahonia duclouxiana. The essential oil was extracted by steam distillation, its chemical components were analyzed by GC-MS, and principal component was confirmed by GC. 80 peaks were detected and 41 components have been identified, which made up more than 90% of total essential oil. The major component was 4-Terpineol (43.73%), the other high content components were alpha-Terpineol (5.23%), (Z)-3-Hexen-1-ol (4.78%), Linalool (4.04%), etc. The chemical components of essential oil of from Mahonia duclouxiana by GC-MS for the first time. This study provided science basis for further research development of Mahonia duclouxiana.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Preise und Verfügbarkeit sind derzeit nicht verfügbar.