Merck
Intrauterine administration of plant oils inhibits luteolysis in the mare.
Equine veterinary journal (2010-12-15)
S Wilsher, W R Allen
ZUSAMMENFASSUNG
The maternal recognition of pregnancy (MRP) signal in the mare has not been determined, although oestrogens have been proposed as a potential candidate. To determine effects of intrauterine administration of oestrogen and various oils on cyclic luteolysis in the mare. Intrauterine oestradiol or fatty acids may suppress luteolysis in the cycling mare when administered during late dioestrus. A single 1 ml dose of slow-release oestradiol (10 mg/ml) in fractionated coconut oil was infused into the uterine lumen of cycling mares on Days 6, 8, 10, 12 or 14 post ovulation (n=12 in each group). Four further groups, each of 12 mares, received an intrauterine infusion of either 1 ml of fractionated coconut oil, peanut oil, mineral oil or a slow-release preparation of oestradiol (10 mg/ml) in mineral oil on Day 10 post ovulation. Serial blood samples were assayed for progesterone concentrations to monitor luteal function. Intrauterine administration of oestradiol in fractionated coconut oil showed peak efficacy at Day 10 when luteolysis was delayed in 11/12 (92%) mares. The ability of the treatment to delay luteolysis was not significantly different when administered on Days 8 (9/12; 75%), 12 (10/12; 83%) or 14 (6/12; 50%) of dioestrus, but declined significantly when given on Day 6 (3/12; 25%). Oestradiol was not needed to initiate luteostasis since fractionated coconut oil alone or peanut oil administered at Day 10 induced the same high rate of luteal persistence (11/12; 92% for both oils). In contrast, mineral oil did not prolong luteal lifespan, either when administered alone (2/12; 17%) or combined with oestradiol (3/12; 25%). These results do not unequivocally rule out a possible involvement of embryonic oestrogens in MRP in the mare but suggest it is unlikely. The results demonstrate that plant oils can postpone luteolysis, suggesting they may modulate synthesis or release of prostaglandins from the mare's endometrium. Administration of fractionated coconut or peanut oil on Day 10 post ovulation provides an effective and practical method of prolonging luteal function ('pseudopregnancy') thereby suppressing unwanted oestrous behaviour. Further studies to elucidate the mechanism by which this is achieved may increase understanding of both luteostasis and MRP signal in the mare.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Erdnussöl, delivery vehicle for lipophilic compounds
Preise und Verfügbarkeit sind derzeit nicht verfügbar.