Merck
Synthesis of (-)-homogalanthamine from naltrexone.
The Journal of organic chemistry (2011-03-09)
Naoshi Yamamoto, Hideaki Fujii, Satomi Imaide, Shigeto Hirayama, Toru Nemoto, Junji Inokoshi, Hiroshi Tomoda, Hiroshi Nagase
ZUSAMMENFASSUNG
Acetylcholinesterase inhibitor (-)-homogalanthamine 3 was synthesized from μ opioid antagonist naltrexone (2) in 16% total yield. The synthesis features Grob fragmentation as a key reaction, which was especially accelerated in the presence of 15-crown-5.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
15-Krone-5, 98%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.