Merck
  • Applicability of sucrose laurate as surfactant in solid dispersions prepared by melt technology.
Applicability of sucrose laurate as surfactant in solid dispersions prepared by melt technology.
International journal of pharmaceutics (2011-03-23)
Angéla Szuts, Péter Láng, Rita Ambrus, Lóránd Kiss, Mária A Deli, Piroska Szabó-Révész
ZUSAMMENFASSUNG
This study focused on an investigation of the applicability of sucrose laurate as surfactant in solid dispersions. Although this surfactant has a US Drug Master File, it has not been used so far in internal pharmaceutical products. High drug-loaded solid dispersion systems consisting of gemfibrozil as a model drug and PEG 6000 as a carrier, with or without sucrose laurate (D1216), were prepared by the melting method. Cytotoxicity studies on Caco-2 monolayer cells were also performed, in order to gain information on the applicability of D1216 in oral formulations. The results showed that the presence of the surface-active agent did not affect the solid-state characteristics of the model drug significantly. A markedly improved dissolution of gemfibrozil from the ternary solid dispersion systems was observed as compared with the binary solid dispersion systems. The optimum concentration range of the D1216 in the formulations was determined to be 5-10%. The effective final concentrations of D1216 in the dissolution experiments proved to be non-toxic towards CaCo-2 cells. The results suggest the potential use of D1216 in innovative internal pharmaceutical formulations.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Saccharose-monolaurat, BioXtra, ≥97.0% (TLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.