Merck
  • Automated quantitative analysis of in-situ NaI measured spectra in the marine environment using a wavelet-based smoothing technique.
Automated quantitative analysis of in-situ NaI measured spectra in the marine environment using a wavelet-based smoothing technique.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine (2011-07-12)
Christos Tsabaris, Aristides Prospathopoulos
ZUSAMMENFASSUNG
An algorithm for automated analysis of in-situ NaI γ-ray spectra in the marine environment is presented. A standard wavelet denoising technique is implemented for obtaining a smoothed spectrum, while the stability of the energy spectrum is achieved by taking advantage of the permanent presence of two energy lines in the marine environment. The automated analysis provides peak detection, net area calculation, energy autocalibration, radionuclide identification and activity calculation. The results of the algorithm performance, presented for two different cases, show that analysis of short-term spectra with poor statistical information is considerably improved and that incorporation of further advancements could allow the use of the algorithm in early-warning marine radioactivity systems.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumiodid, ACS reagent, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumiodid, ReagentPlus®, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumiodid, 99.999% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumiodid, ≥99.99% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumiodid, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.