Merck
  • Trimethylsilyldiazomethane derivatization coupled with solid-phase extraction for the determination of alendronate in human plasma by LC-MS/MS.
Trimethylsilyldiazomethane derivatization coupled with solid-phase extraction for the determination of alendronate in human plasma by LC-MS/MS.
Analytical and bioanalytical chemistry (2011-10-18)
Meixia Chen, Ke Liu, Dafang Zhong, Xiaoyan Chen
ZUSAMMENFASSUNG
Alendronate is an important representative of bisphosphonates, strongly polar compounds that lack chromophores. With rare exceptions, derivatization of the analytes is necessary for bioanalysis. In this study, a rapid liquid chromatography-tandem mass spectrometry method employing pre-column derivatization was developed and validated for the determination of alendronate concentrations in human plasma. The procedure was based on derivatization with trimethylsilyldiazomethane during solid-phase extraction on a weak anion-exchange solid-phase cartridge, which integrated sample purification and derivatization into one step. The alendronate derivative was eluted with methanol. Chromatographic separation was performed on a Capcell PAK-C18 column. The total run time was 6.5 min. The calibration curve was linear in the range 1.00-1,000 ng/mL using d6-alendronate as the internal standard. The lower limit of quantification was 1.00 ng/mL. The intra- and inter-assay precision (in RSD) calculated from quality control samples was less than 15%, and the accuracy was between 98.1% and 100.2%. The validated method was successfully applied to characterize the pharmacokinetic profiles of alendronate following the intravenous infusion of 5 or 10 mg alendronate sodium to healthy volunteers.