- Functionalized 2,3-dihydrofurans via palladium-catalyzed oxyarylation of α-allyl-β-ketoesters.
Functionalized 2,3-dihydrofurans via palladium-catalyzed oxyarylation of α-allyl-β-ketoesters.
Organic & biomolecular chemistry (2011-11-02)
Sandro Cacchi, Giancarlo Fabrizi, Antonella Goggiamani, Antonia Iazzetti, David Madec, Giovanni Poli, Guillaume Prestat
PMID22041862
ZUSAMMENFASSUNG
The palladium-catalyzed reaction of (hetero)aryl bromides, chlorides, and nonaflates with α-allyl-β-ketoesters provides ready efficient access to functionalized 2,3-dihydrofurans. The reaction tolerates several useful substituents including chloro, fluoro, ether, ketone, ester, cyano, and nitro groups.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Preise und Verfügbarkeit sind derzeit nicht verfügbar.