Merck
  • Enzymatic coproduction of biodiesel and glycerol carbonate from soybean oil and dimethyl carbonate.
Enzymatic coproduction of biodiesel and glycerol carbonate from soybean oil and dimethyl carbonate.
Enzyme and microbial technology (2011-11-25)
Pil-Je Seong, Byoung Wook Jeon, Myunggu Lee, Dae Haeng Cho, Duk-Ki Kim, Kwang S Jung, Seung Wook Kim, Sung Ok Han, Yong Hwan Kim, Chulhwan Park
ZUSAMMENFASSUNG
The enzymatic coproduction of biodiesel and glycerol carbonate by the transesterification of soybean oil was studied using lipase as catalyst in organic solvent. To produce biodiesel and glycerol carbonate simultaneously, experiments were designed sequentially. Enzyme screening, the molar ratio of dimethyl carbonate (DMC) to soybean oil, reaction temperature and solvent effects were investigated. The results of enzyme screening, at 100 g/L Novozym 435 (immobilized Candida antarctica lipase B), biodiesel and glycerol carbonate showed conversions of 58.7% and 50.7%, respectively. The optimal conditions were 60 °C, 100 g/L Novozym 435, 6.0:1 molar ratio with tert-butanol as solvent: 84.9% biodiesel and 92.0% glycerol carbonate production was achieved.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylcarbonat, anhydrous, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylcarbonat, ReagentPlus®, 99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.