Merck
  • Strain-dependent effects of the histamine H₄ receptor antagonist JNJ7777120 in a murine model of acute skin inflammation.
Strain-dependent effects of the histamine H₄ receptor antagonist JNJ7777120 in a murine model of acute skin inflammation.
Experimental dermatology (2011-12-14)
Gabriella Coruzzi, Cristina Pozzoli, Maristella Adami, Daniela Grandi, Nicoletta Guido, Rogier Smits, Iwan de Esch, Rob Leurs
ZUSAMMENFASSUNG
The effects of the histamine H(4) receptor antagonist JNJ7777120 were evaluated in a model of acute skin inflammation induced by local application of croton oil. The influence of strain on the effect of JNJ7777120 was investigated in four different mouse strains (CD-1, NMRI, BALB/c and C57BL/6J). In CD-1 mice, JNJ777720 (30-100 mg/kg subcutaneously, s.c.) exerted a dose-dependent inhibition of croton oil-induced ear inflammation and polymorphonuclear leucocyte infiltration, as confirmed by histological evaluation of ear tissues. JNJ7777120 (30-100 mg/kg) did not reduce ear oedema in NMRI, BALB/c or C57BL/6J mice. The positive control, dexamethasone (2 mg/kg s.c.) induced significant anti-inflammatory effects only in CD-1 and NMRI mice. In these strains, also the histamine H(1) -receptor blocker pyrilamine (30 mg/kg s.c.) significantly reduced ear oedema at 2 h after croton oil challenge, being as effective as JNJ7777120 in CD-1 mice. Taken together, these data demonstrate that the H(4) receptor antagonist JNJ7777120 may reduce acute croton oil-induced skin inflammation as effectively as H(1) receptor blockade. However, present experiments evidenced for the first time marked strain-related differences in the JNJ7777120 pharmacological activity, which have to be carefully considered when using this ligand to characterize histamine H(4) receptor functions in murine models and translating preclinical data to clinical human settings.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Crotonöl
Preise und Verfügbarkeit sind derzeit nicht verfügbar.