- [Histrelin acetate--the first once yearly LHRH agonist].
[Histrelin acetate--the first once yearly LHRH agonist].
Lijecnicki vjesnik (2011-12-15)
Silvio Altarac
PMID22165080
ZUSAMMENFASSUNG
Long-acting synthetic luteinising hormone-releasing hormone agonists have become the mainstay for androgen-deprivation therapy, because they avoid the physical and psychological discomfort associated with orchidectomy and lack the potential cardiotoxicity associated with estrogens such as diethylstilbestrol. Currently available luteinising hormone-releasing hormone agonist analogues include leuprolide, goserelin, triptorelin, degarelix and buserelin were administered as either intramuscular or subcutaneous depot injections on a 1, 2, 3 or 6 months basis. Histrelin acetate is the first long-acting luteinising hormone-releasing hormone agonist available as a once-yearly subcutaneous implant.