Merck
  • Distinct receptor subtypes mediate arginine vasopressin-dependent ACTH release and intracellular calcium mobilization in rat pituitary cells.
Distinct receptor subtypes mediate arginine vasopressin-dependent ACTH release and intracellular calcium mobilization in rat pituitary cells.
European journal of pharmacology (2012-01-31)
Elisabetta Perdonà, Roberto Arban, Cristiana Griffante
ZUSAMMENFASSUNG
In the present study, adrenocorticotropic hormone (ACTH) release and intracellular calcium ([Ca(2+)](i)) increase induced by arginine vasopressin (AVP) were characterized in collagenase-dispersed and 3-day cultured rat anterior pituitary cells. AVP and the selective vasopressin V(1b) receptor agonist, [1-deamino-4-cyclohexylalanine]AVP (d[Cha(4)]AVP) induced ACTH release with nanomolar potencies in both cell preparations, and produced a maximal stimulation that was about 1.5 fold greater in the 3-day cultured cells, indicating that the vasopressin V(1b) receptor-ACTH release pathway is enhanced over time in culture. In dispersed cells, AVP, oxytocin and d[Cha(4)]AVP induced [Ca(2+)](i) increases with nanomolar potencies. The selective vasopressin V(1a) receptors antagonist, SR49059 (100 nM), together with the selective oxytocin receptors antagonist (d(CH(2))(5)(1)Tyr(Me)(2),Thr(4),Orn(8),Tyr-NH(2)(9)-vasotocin (100 nM), inhibited the maximal AVP response by ~70%, without affecting the response to d[Cha(4)]AVP, suggesting that the V(1b) receptor was only partially responsible for the AVP-induced [Ca(2+)](i) increase. In contrast, in 3-day cultures, AVP induced an increase in [Ca(2+)](i), while oxytocin and d[Cha(4)]AVP did not. The response to AVP was completely antagonized by SR49059, whereas the vasopressin V(1b) receptor antagonists, SSR149415 and (d(CH(2))(5)(1)Tyr(Me)(2),Thr(4),Orn(8),Tyr-NH(2)(9))-vasotocin had no effect, indicating that the [Ca(2+)](i) increase was mediated exclusively by vasopressin V(1a) receptors. In conclusion, the enhancement of vasopressin V(1b) receptor-mediated ACTH release and the lack of a detectable vasopressin V(1b) receptor coupling to [Ca(2+)](i) increase in cultured cells suggests the activation of a different/additional signaling pathway in the molecular mechanism of ACTH release.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
SR 49059, ≥97% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.