Merck
Renewable cathode materials from biopolymer/conjugated polymer interpenetrating networks.
Science (New York, N.Y.) (2012-03-24)
Grzegorz Milczarek, Olle Inganäs
ZUSAMMENFASSUNG
Renewable and cheap materials in electrodes could meet the need for low-cost, intermittent electrical energy storage in a renewable energy system if sufficient charge density is obtained. Brown liquor, the waste product from paper processing, contains lignin derivatives. Polymer cathodes can be prepared by electrochemical oxidation of pyrrole to polypyrrole in solutions of lignin derivatives. The quinone group in lignin is used for electron and proton storage and exchange during redox cycling, thus combining charge storage in lignin and polypyrrole in an interpenetrating polypyrrole/lignin composite.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lignosulfonsäure Natriumsalz, average Mw ~52,000, average Mn ~7,000
Preise und Verfügbarkeit sind derzeit nicht verfügbar.