Merck
  • Anti-inflammatory effects of a polyphenols-rich extract from tea (Camellia sinensis) flowers in acute and chronic mice models.
Anti-inflammatory effects of a polyphenols-rich extract from tea (Camellia sinensis) flowers in acute and chronic mice models.
Oxidative medicine and cellular longevity (2012-08-18)
Bang-Tian Chen, Wei-Xi Li, Rong-Rong He, Yi-Fang Li, Bun Tsoi, Yu-Jia Zhai, Hiroshi Kurihara
ZUSAMMENFASSUNG
While beneficial health properties of tea leaves have been extensively studied, less attention is paid to the flowers of tea. In this study, the anti-inflammatory effects of hot water extract of tea (Camellia sinensis) flowers were investigated. Pharmacological studies found that administration of tea flowers extract (TFE) could effectively inhibit croton oil-induced ear edema and carrageenin-induced paw edema. Furthermore, administration of TFE also protected against Propionibacterium acnes (P. ances) plus lipopolysaccharide-(LPS-) induced liver inflammation by reversing the histologic damage and plasma alanine aminotransferase (ALT) increase. Moreover, the levels of nitric oxide (NO), tumor necrosis factor-(TNF)-α and interleukin-(IL-) 1β mRNA in mouse liver were markedly suppressed after treatment with TFE in mice with immunological liver inflammation. These results indicated that tea flowers had potent anti-inflammatory effects on acute and immunological inflammation in vivo, and may be used as a functional natural food.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Crotonöl
Preise und Verfügbarkeit sind derzeit nicht verfügbar.