Merck
  • Protective effects of sinapic acid on lysosomal dysfunction in isoproterenol induced myocardial infarcted rats.
Protective effects of sinapic acid on lysosomal dysfunction in isoproterenol induced myocardial infarcted rats.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2012-08-28)
Subhro Jyoti Roy, Ponnian Stanely Mainzen Prince
ZUSAMMENFASSUNG
In the pathology of myocardial infarction, lysosomal lipid peroxidation and resulting enzyme release play an important role. We evaluated the protective effects of sinapic acid on lysosomal dysfunction in isoproterenol induced myocardial infarcted rats. Male Wistar rats were treated with sinapic acid (12 mg/kg body weight) orally daily for 10 days and isoproterenol (100 mg/kg body weight) was injected twice at an interval of 24 h (9th and 10th day). Then, lysosomal lipid peroxidation, lysosomal enzymes in serum, heart homogenate, lysosomal fraction and myocardial infarct size were measured. Isoproterenol induced myocardial infarcted rats showed a significant increase in serum creatine kinase-MB and lysosomal lipid peroxidation. The activities of β-glucuronidase, β-galactosidase, cathepsin-B and D were significantly increased in serum, heart and the activities of β-glucuronidase and cathepsin-D were significantly decreased in lysosomal fraction of myocardial infarcted rats. Pre-and-co-treatment with sinapic acid normalized all the biochemical parameters and reduced myocardial infarct size in myocardial infarcted rats. In vitro studies confirmed the free radical scavenging effects of sinapic acid. The possible mechanisms for the observed effects are attributed to sinapic acid's free radical scavenging and membrane stabilizing properties. Thus, sinapic acid has protective effects on lysosomal dysfunction in isoproterenol induced myocardial infarcted rats.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Sinapinsäure, ≥98%, powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Sinapinsäure, suitable for matrix substance for MALDI-MS, ≥99.0% (T)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Sinapinsäure, suitable for matrix substance for MALDI-MS, ≥99.5%, Ultra pure
Preise und Verfügbarkeit sind derzeit nicht verfügbar.