Merck
  • Evaluation of joint toxicity of nitroaromatic compounds and copper to Photobacterium phosphoreum and QSAR analysis.
Evaluation of joint toxicity of nitroaromatic compounds and copper to Photobacterium phosphoreum and QSAR analysis.
Journal of hazardous materials (2012-10-24)
Limin Su, Xujia Zhang, Xing Yuan, Yuanhui Zhao, Dongmei Zhang, Weichao Qin
ZUSAMMENFASSUNG
The individual toxicities of Cu and 11 nitroaromatic compounds to Photobacterium phosphoreum were determined. The toxicity was expressed as the concentrations causing a 50% inhibition of bioluminescence after 15 min exposure (IC(50)). To evaluate the joint effect between the metal ion and the 11 nitroaromatic compounds, the joint toxicity of Cu and 11 nitroaromatic compounds were measured at different Cu concentrations (0.2IC(50), 0.5IC(50) and 0.8IC(50)), respectively. The result shows that the binary joint effect between Cu and nitroaromatic compounds is mainly simple addition at the low Cu concentration (0.2IC(50)). However, an antagonism effect, 55% and 64%, was observed between Cu and 11 nitroaromatic compounds for Cu at medium and high concentrations (0.5IC(50) and 0.8IC(50)). Quantitative structure-activity relationship (QSAR) analysis was performed to study the joint toxicity for the 11 nitroaromatic compounds. The result shows that the toxicity of nitroaromatic compounds is related to descriptors of Connolly solvent-excluded volume (CSEV) and dipolarity/polarizability (S) at low Cu concentration. On the other hand, the toxicity is related to Connolly accessible area (CAA) at medium and high Cu concentrations. The result indicates that different QSAR models on complex mixtures need to be developed to assess the ecological risk in real environments. Using single toxic data to evaluate the toxic effect of mixtures may result in wrong conclusions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kupfer(II)-sulfat, ReagentPlus®, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Kupfer(II)-Sulfat -Lösung, 4 % (w/v) (prepared from copper (II) sulfate pentahydrate)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Kupfer(II)-sulfat, anhydrous, powder, ≥99.99% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.