Merck
  • Effect of menatetrenone, a vitamin k2 analog, on recurrence of hepatocellular carcinoma after surgical resection: a prospective randomized controlled trial.
Effect of menatetrenone, a vitamin k2 analog, on recurrence of hepatocellular carcinoma after surgical resection: a prospective randomized controlled trial.
Anticancer research (2012-12-12)
Mitsuru Ishizuka, Keiichi Kubota, Mitsugi Shimoda, Junji Kita, Masato Kato, Kyung Hwa Park, Takayuki Shiraki
ZUSAMMENFASSUNG
The aim of this study was to investigate whether menatetrenone (MNT) suppresses hepatocellular carcinoma (HCC) recurrence in patients undergoing hepatectomy. Between January 2005 and September 2009, 101 patients who underwent curative hepatectomy for primary HCC were enrolled in the study. Patients were divided into two groups: a non-MNT group (n=51), and an MNT group (n=50) that was administered 45 mg of MNT daily. During the observation period, recurrence was observed in 33 patients in the non-MNT group and in 28 patients of the MNT group (p=0.545). In patients with a preoperative Des-γ-carboxy-prothrombin (DCP) level lower than 40 AU/l (n=38), the cumulative disease-free survival rates at 12, 36, and 60 months in the non-MNT group, were 81.3%, 0.0%, and 0.0%, respectively, while those in the MNT group were 78.3%, 58.1%, and 31.0%, respectively (p=0.060). MNT has a moderately suppressive effect on HCC recurrence after hepatectomy, especially in patients with a normal preoperative DCP level.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Vitamin K2
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Menachinon (K2), analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.