Merck
  • Wogonin induced calreticulin/annexin A1 exposure dictates the immunogenicity of cancer cells in a PERK/AKT dependent manner.
Wogonin induced calreticulin/annexin A1 exposure dictates the immunogenicity of cancer cells in a PERK/AKT dependent manner.
PloS one (2012-12-20)
Yong Yang, Xian-Jing Li, Zhen Chen, Xuan-Xuan Zhu, Jing Wang, Lin-bo Zhang, Lei Qiang, Yan-Jun Ma, Zhi-yu Li, Qing-Long Guo, Qi-Dong You
ZUSAMMENFASSUNG
In response to ionizing irradiation and certain chemotherapeutic agents, dying tumor cells elicit a potent anticancer immune response. However, the potential effect of wogonin (5,7-dihydroxy-8-methoxyflavone) on cancer immunogenicity has not been studied. Here we demonstrated for the first time that wogonin elicits a potent antitumor immunity effect by inducing the translocation of calreticulin (CRT) and Annexin A1 to cell plasma membrane as well as the release of high-mobility group protein 1 (HMGB1) and ATP. Signal pathways involved in this process were studied. We found that wogonin-induced reactive oxygen species (ROS) production causes an endoplasmic reticulum (ER) stress response, including the phosphorylation of PERK (PKR-like endoplasmic reticulum kinase)/PKR (protein kinase R) and eIF2α (eukaryotic initiation factor 2α), which served as upstream signal for the activation of phosphoinositide 3-kinase (PI3K)/AKT, inducing calreticulin (CRT)/Annexin A1 cell membrane translocation. P22/CHP, a Ca(2+)-binding protein, was associated with CRT and was required for CRT translocation to cell membrane. The releases of HMGB1 and ATP from wogonin treated MFC cells, alone or together with other possible factors, activated dendritic cells and induced cytokine releases. In vivo study confirmed that immunization with wogonin-pretreated tumor cells vaccination significantly inhibited homoplastic grafted gastric tumor growth in mice and a possible inflammatory response was involved. In conclusion, the activation of PI3K pathway elicited by ER stress induced CRT/Annexin A1 translocation ("eat me" signal) and HMGB1 release, mediating wogonin-induced immunity of tumor cell vaccine. This indicated that wogonin is a novel effective candidate of immunotherapy against gastric tumor.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Rapamycin, ≥95% (HPLC), powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
LY 294002, LY294002, CAS 154447-36-6, is a cell-permeable, potent, reversible, and specific inhibitor of PI 3-kinase ((IC50 = 1.4 µM). Acts on the ATP-binding site.
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Wogonin hydrate, ≥98% (HPLC), solid
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Akt-Inhibitor X, The Akt Inhibitor X, also referenced under CAS 925681-41-0, controls the biological activity of Akt. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Preise und Verfügbarkeit sind derzeit nicht verfügbar.