Merck
  • Immunomodulatory effect of a proanthocyanidin-rich extract from Pinus radiata bark by dosing period in chickens.
Immunomodulatory effect of a proanthocyanidin-rich extract from Pinus radiata bark by dosing period in chickens.
Poultry science (2013-01-10)
In-Jae Park, Se-Yeoun Cha, Min Kang, Hyung-Kwan Jang
ZUSAMMENFASSUNG
We evaluated the immunomodulatory effects of a proanthocyanidin-rich extract (PAE) from Pinus radiata bark in Korean native chickens. In experiment 1, proliferation of peripheral blood mononuclear cells was significantly enhanced in chickens administered 1.25, 2.5, 5, and 10 mg/kg of PAE for 2 wk once daily by oral gavage compared with no administration of PAE in vitro. Proliferation of splenocyte and thymocyte cells was also significantly enhanced in chickens administered 1.25, 2.5, 5, and 10 mg/kg of PAE for 5 wk. In experiment 2, proliferation of peripheral blood mononuclear cells, splenocytes, and thymocytes in chickens administered PAE for 6 wk was continuously higher than that in other groups. Additionally, increasing cell populations for which CD4(+)CD8(-)(Th cells) and Bu-1(+) (B cells) protein was expressed on the cell surface were significantly different after 4 wk of PAE administration in chickens. Thus, the immunomodulatory effects of a PAE from P. radiata bark differed by dosing period in chickens.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Concanavalin A aus Canavalia ensiformis (Jack bean), Type IV-S, lyophilized powder, aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Concanavalin A aus Canavalia ensiformis (Jack bean), Type IV, lyophilized powder
Sigma-Aldrich
Concanavalin A aus Canavalia ensiformis (Jack bean), Type VI, lyophilized powder
Sigma-Aldrich
Concanavalin A aus Canavalia ensiformis (Jack bean), Type IV-S, lyophilized powder, γ-irradiated, BioReagent, suitable for cell culture