Merck
Elucidating the regulon of multidrug resistance regulator RarA in Klebsiella pneumoniae.
Antimicrobial agents and chemotherapy (2013-01-16)
Shyamasree De Majumdar, Mark Veleba, Sarah Finn, Séamus Fanning, Thamarai Schneiders
ZUSAMMENFASSUNG
RarA is an AraC-type regulator in Klebsiella pneumoniae, which, when overexpressed, confers a low-level multidrug-resistant (MDR) phenotype linked to the upregulation of both the acrAB and oqxAB efflux genes. Increased rarA expression has also been shown to be integral in the development of tigecycline resistance in the absence of ramA in K. pneumoniae. Given its phenotypic role in MDR, microarray analyses were performed to determine the RarA regulon. Transcriptome analysis was undertaken using strains Ecl8ΔrarA/pACrarA-2 (rarA-expressing construct) and Ecl8ΔrarA/pACYC184 (vector-only control) using bespoke microarray slides consisting of probes derived from the genomic sequences of K. pneumoniae MGH 78578 (NC_009648.1) and Kp342 (NC_011283.1). Our results show that rarA overexpression resulted in the differential expression of 66 genes (42 upregulated and 24 downregulated). Under the COG (clusters of orthologous groups) functional classification, the majority of affected genes belonged to the category of cell envelope biogenesis and posttranslational modification, along with genes encoding the previously uncharacterized transport proteins (e.g., KPN_03141, sdaCB, and leuE) and the porin OmpF. However, genes associated with energy production and conversion and amino acid transport/metabolism (e.g., nuoA, narJ, and proWX) were found to be downregulated. Biolog phenotype analyses demonstrated that rarA overexpression confers enhanced growth of the overexpresser in the presence of several antibiotic classes (i.e., beta-lactams and fluoroquinolones), the antifungal/antiprotozoal compound clioquinol, disinfectants (8-hydroxyquinoline), protein synthesis inhibitors (i.e., minocycline and puromycin), membrane biogenesis agents (polymyxin B and amitriptyline), DNA synthesis (furaltadone), and the cytokinesis inhibitor (sanguinarine). Both our transcriptome and phenotypic microarray data support and extend the role of RarA in the MDR phenotype of K. pneumoniae.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Puromycin -dihydrochlorid aus Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Puromycin -dihydrochlorid, Ready Made Solution, from Streptomyces alboniger, 10 mg/mL in H2O, suitable for cell culture
Sigma-Aldrich
Polymyxin B -sulfat (Salz), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Puromycin -dihydrochlorid aus Streptomyces alboniger, ≥98% (HPLC), powder
Sigma-Aldrich
Polymyxin B -sulfat (Salz)
Sigma-Aldrich
Amitriptylin -hydrochlorid, ≥98% (TLC), powder
Sigma-Aldrich
Polymyxin B -sulfat, meets USP testing specifications
Sigma-Aldrich
Polymyxin B -Lösung, 20 mg/mL in H2O
Supelco
Amitriptyline hydrochloride solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Polymyxin B -Lösung, 1 mg/mL in H2O, analytical standard
Sigma-Aldrich
Furaltadon
Supelco
Furaltadon, VETRANAL®, analytical standard
Polymyxin-B-sulfat, European Pharmacopoeia (EP) Reference Standard