Merck
  • Perfluoroalkylated compounds induce cell death and formation of reactive oxygen species in cultured cerebellar granule cells.
Perfluoroalkylated compounds induce cell death and formation of reactive oxygen species in cultured cerebellar granule cells.
Toxicology letters (2013-01-24)
Trine Reistad, Frode Fonnum, Espen Mariussen
ZUSAMMENFASSUNG
The present communication investigates the effects of different perfluoroalkylated compounds (PFCs) on formation of reactive oxygen species (ROS) and cell death in cultured cerebellar granule cells. This allows direct comparison with similar effects found for other environmental contaminants like polychlorinated biphenyls and brominated flame-retardants. The increase in ROS formation and cell death was assayed using the fluorescent probe 2,7-dichlorofluorescin diacetate (DCFH-DA) and the trypan blue exclusion assay. The effects of the PFCs were structure dependent. Cell death was induced at relatively low concentrations by perfluorooctyl sulfonate (PFOS), perfluorooctane sulfonylamide (PFOSA) and the fluorotelomer alcohol 1H, 1H, 2H, 2H-perfluorodecanol (FTOH 8:2) with EC(50)-values of 62 ± 7.6, 13 ± 1.8 and 15 ± 4.2 μM (mean ± SD) respectively. PFOS, perfluorooctanoic acid (PFOA) and PFOSA induced a concentration dependent increase in ROS formation with EC(50)-values of 27 ± 9.0, 25 ± 11 and 57 ± 19μM respectively. Reduced cell viability and ROS formation were observed at concentration level close to what is found in serum of occupationally exposed workers. The effect of PFCs on ROS formation and cell viability was compared with other halogenated compounds and future investigations should emphasize effects of mixtures and how physical chemical properties of the compounds influence their toxicity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Perfluoroctansäure, 95%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Heptadecafluoroctansulfonsäure -Lösung, 100 μg/mL in methanol, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Pentadecafluoroctansäure Ammoniumsalz, ≥98.0% (sum of isomers)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Pentadecafluoroctansäure -Lösung, 100 μg/mL in methanol, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.