Merck
  • Molecular cloning, recombinant expression and characterization of GMCSF from the rhesus monkey, Macaca mulatta.
Molecular cloning, recombinant expression and characterization of GMCSF from the rhesus monkey, Macaca mulatta.
Developmental and comparative immunology (2013-01-29)
Ze Tao, Hao Yang, Dianlong Jia, Lin Wan, Jingqiu Cheng, Xiaofeng Lu
ZUSAMMENFASSUNG
Recent studies have found that, in addition to hematopoiesis, granulocyte-macrophage colony-stimulating factor (GMCSF) plays pivotal roles in multiple immune disorders. The gene encoding Macaca mulatta GMCSF (mmGMCSF) was cloned from stimulated peripheral blood mononuclear cells (PBMCs). Concanavalin A (Con A) and mismatched allogeneic antigen-stimulation significantly increased the production of mmGMCSF by monkey PBMCs. The gene encoding mature mmGMCSF was first expressed as a soluble fusion protein in Escherichia coli, and native mmGMCSF was further prepared by protease cleavage. The recombinant mmGMCSF induced antigen-presenting dendritic cells from monkey PBMCs, suggesting a central role of mmGMCSF in the immune system of the rhesus monkey. Although the predicted mature mmGMCSF protein differs from human GMCSF (hGMCSF) at six amino acid residues, mmGMCSF showed a strong ability to support human TF-1 cell survival. Additional co-immunoprecipitation experiments revealed that mmGMCSF cross reacts with the hGMCSF receptor (hGMCSFR). In addition, the hGMCSF-neutralizing agents hGMCSFR-Fc and anti-hGMCSF antibody reduced the biological effects of mmGMCSF on TF-1 cells. These results suggest that recombinant mmGMCSF might be used for the in vitro evaluation of novel hGMCSF-neutralizing agents prior to the in vivo preclinical evaluation of these agents in the rhesus monkey model.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Concanavalin A aus Canavalia ensiformis (Jack bean), Type IV-S, lyophilized powder, aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Concanavalin A aus Canavalia ensiformis (Jack bean), Type IV, lyophilized powder
Sigma-Aldrich
Concanavalin A aus Canavalia ensiformis (Jack bean), Type VI, lyophilized powder
Sigma-Aldrich
Concanavalin A aus Canavalia ensiformis (Jack bean), Type IV-S, lyophilized powder, γ-irradiated, BioReagent, suitable for cell culture