- [Pyomyositis in children: report of two cases].
[Pyomyositis in children: report of two cases].
Revista chilena de infectologia : organo oficial de la Sociedad Chilena de Infectologia (2013-03-02)
Felipe Cavagnaro, Jaime Rodríguez, M Eugenia Arancibia, Bárbara Walker, Aníbal Espinoza
PMID23450416
ZUSAMMENFASSUNG
Pyomyositis (PM) is an uncommon pyogenic infection of skeletal muscle and, when not properly treated, it can progress to a high-risk clinical situation with high mortality. Because it usually has a subacute presentation, diagnosis is often delayed. We present two cases of PM of the paraspinal muscles in healthy children and discuss the current state of knowledge of this disease.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Preise und Verfügbarkeit sind derzeit nicht verfügbar.