Merck
  • The morphology and fabrication of nanostructured micelle by a novel block copolymer with linear-dendritic structure.
The morphology and fabrication of nanostructured micelle by a novel block copolymer with linear-dendritic structure.
Materials science & engineering. C, Materials for biological applications (2013-03-19)
Rulin Cai, Rui Li, Jiasheng Qian, Anjian Xie, Kangming Nie
ZUSAMMENFASSUNG
We report here a novel approach to fabricate a nanostructured micelle as potential drug carriers and the relationship between the morphological structure and the preparation condition. The polymeric micelle aggregates constructed by self-assembly compose of the poly(ε-caprolactone)/monomethoxy poly(ethylene glycol) linear-dendritic block copolymers. The corresponding copolymers were synthesized by using ring opening polymerization of ε-caprolactone (CL) and a dendritic poly(ether-amide) (DPEA-OH) as an initiator, then coupling with the isocyanate end-capped linear monomethoxy polyethylene glycol. Fluorescence spectroscopy, dynamic light scattering (DLS) and transmission electron microscopy (TEM) were performed to characterize the copolymer micelles. The critical micelle concentration (CMC) was determined to be 1.623 mg/L. The hydrodynamic radius (Rh) and the polydispersity index (PDI) are influenced by the concentration of the micelle solutions. The multiple morphologies of the micelle aggregates, including spheres, rob-like dendritic structure and vesicles were observed, which the variety depends on the various preparation conditions. The nanostructured micelles based on the linear-dendritic block copolymer possess the strong thermodynamic stability and the power of solubilization of hydrophobic drug molecules.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Polycaprolacton, average Mn 80,000
Sigma-Aldrich
Benzoesäure, ACS reagent, ≥99.5%
Supelco
Benzoesäure, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Polycaprolacton, average Mw ~14,000, average Mn ~10,000 by GPC
Sigma-Aldrich
Pyren, 98%
Sigma-Aldrich
Poly(ethylenglycol)methylether, average Mn 5,000
Sigma-Aldrich
Polycaprolacton, average Mn 45,000
Supelco
Mettler-Toledo Kalibriersubstanz ME 18555, Benzoesäure, analytical standard, (for the calibration of the melting point system), traceable to primary standards (LGC)
Sigma-Aldrich
Poly(ethylenglycol)methylether, average Mn ~2,000
Sigma-Aldrich
Poly(ethylenglycol)methylether, average Mn 550
Sigma-Aldrich
Polyethylenglykol 350-monomethylether, average mol wt 350
Supelco
Schmelzpunktstandard 121-123°C, analytical standard
Sigma-Aldrich
Benzoesäure, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
Sigma-Aldrich
Poly(ethylenglycol)methylether, average Mn 750
Supelco
Benzoesäure, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Pyren, puriss. p.a., suitable for fluorescence, ≥99.0% (GC)
Supelco
Pyren, analytical standard
Sigma-Aldrich
Benzoesäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoesäure, ReagentPlus®, 99%
Sigma-Aldrich
Poly(ethylenglycol)methylether, average Mn 10,000
Sigma-Aldrich
Benzoesäure, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
Sigma-Aldrich
Poly(ethylenglycol)methylether, BioUltra, 500
Supelco
Benzoesäure, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Pyren, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Benzoesäure, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Kaliumbenzoat, ReagentPlus®, 99%
Sigma-Aldrich
Poly(ethylenglycol)methylether, average Mn 20,000
Sigma-Aldrich
Benzoesäure, purified by sublimation, ≥99%
Sigma-Aldrich
Pyren, sublimed grade, 99%
Pyren, BCR®, certified reference material