Merck
  • Prophylactic oral/topical non-absorbed antifungal agents to prevent invasive fungal infection in very low birth weight infants.
Prophylactic oral/topical non-absorbed antifungal agents to prevent invasive fungal infection in very low birth weight infants.
The Cochrane database of systematic reviews (2013-04-02)
Nicola Austin, Brian A Darlow, William McGuire
ZUSAMMENFASSUNG
Invasive fungal infection is an important cause of mortality and morbidity in very preterm or very low birth weight infants. Uncertainty exists about the effect of prophylactic oral/topical non-absorbed antifungals to reduce mucocutaneous colonisation and so limit the risk of invasive fungal infection in this population. To assess the effect of prophylactic oral/topical non-absorbed antifungal therapy on the incidence of invasive fungal infection, mortality and morbidity in very preterm or very low birth weight infants. We used the standard search strategy of the Cochrane Neonatal Review Group. This included searches of the Cochrane Central Register of Controlled Trials (CENTRAL: The Cochrane Library, 2012, Issue 3), MEDLINE, EMBASE, and CINAHL (to August 2012), conference proceedings, and previous reviews. Randomised controlled trials or quasi-randomised controlled trials that compared the effect of prophylactic oral/topical non-absorbed antifungal therapy versus placebo or no drug or another antifungal agent or dose regimen in very preterm or very low birth weight infants. We extracted data using the standard methods of the Cochrane Neonatal Review Group with separate evaluation of trial quality and data extraction by two review authors. Four trials, in which a total of 1800 infants participated, compared oral/topical non-absorbed antifungal prophylaxis (nystatin or miconazole) with placebo or no drug. These trials had various methodological weaknesses including quasi-randomisation, lack of allocation concealment, and lack of blinding of intervention and outcomes assessment. The incidence of invasive fungal infection was very high in the control groups of three of these trials. Meta-analysis found a statistically significant reduction in the incidence of invasive fungal infection [typical risk ratio 0.20 (95% confidence interval 0.14 to 0.27); risk difference -0.18 (-0.21 to -0.16)] but substantial statistical heterogeneity was present. We did not find a statistically significant effect on mortality [typical risk ratio 0.87 (0.72 to 1.05); risk difference -0.03 (-0.06 to 0.01)]. None of the trials assessed posthospital discharge outcomes.Two trials (N = 265) assessed the effect of oral/topical non-absorbed versus systemic antifungal prophylaxis. Meta-analyses did not find any statistically significant differences in the incidences of invasive fungal infection or all-cause mortality. The finding of a reduction in risk of invasive fungal infection in very low birth weight infants treated with oral/topical non-absorbed antifungal prophylaxis should be interpreted cautiously because of methodological weaknesses in the included trials. Further large randomised controlled trials in current neonatal practice settings are needed to resolve this uncertainty. These trials might compare oral/topical non-absorbed antifungal agents with placebo, with each other, or with systemic antifungal agents and should include an assessment of effect on long-term neurodevelopmental outcomes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Nystatin-Suspension, suspension, 10,000 unit/mL in DPBS, BioReagent, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Nystatin, powder, BioReagent, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Fluconazol, ≥98% (HPLC), powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Nystatin, ≥4,400 units/mg
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Fluconazol, Pharmaceutical Secondary Standard; Certified Reference Material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Nystatin, powder, γ-irradiated, BioXtra, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
(±)-Miconazol -nitrat (Salz), imidazole antibiotic
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
(±)-Miconazol -nitrat (Salz), Pharmaceutical Secondary Standard; Certified Reference Material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Nystatin, European Pharmacopoeia (EP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Fluconazol, European Pharmacopoeia (EP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Miconazolnitrat, European Pharmacopoeia (EP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Fluconazol für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Miconazol, European Pharmacopoeia (EP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.