Merck
  • RecJ-like protein from Pyrococcus furiosus has 3'-5' exonuclease activity on RNA: implications for proofreading of 3'-mismatched RNA primers in DNA replication.
RecJ-like protein from Pyrococcus furiosus has 3'-5' exonuclease activity on RNA: implications for proofreading of 3'-mismatched RNA primers in DNA replication.
Nucleic acids research (2013-04-23)
Hui Yuan, Xi-Peng Liu, Zhong Han, Thorsten Allers, Jing-Li Hou, Jian-Hua Liu
ZUSAMMENFASSUNG
Replicative DNA polymerases require an RNA primer for leading and lagging strand DNA synthesis, and primase is responsible for the de novo synthesis of this RNA primer. However, the archaeal primase from Pyrococcus furiosus (Pfu) frequently incorporates mismatched nucleoside monophosphate, which stops RNA synthesis. Pfu DNA polymerase (PolB) cannot elongate the resulting 3'-mismatched RNA primer because it cannot remove the 3'-mismatched ribonucleotide. This study demonstrates the potential role of a RecJ-like protein from P. furiosus (PfRecJ) in proofreading 3'-mismatched ribonucleotides. PfRecJ hydrolyzes single-stranded RNA and the RNA strand of RNA/DNA hybrids in the 3'-5' direction, and the kinetic parameters (Km and Kcat) of PfRecJ during RNA strand digestion are consistent with a role in proofreading 3'-mismatched RNA primers. Replication protein A, the single-stranded DNA-binding protein, stimulates the removal of 3'-mismatched ribonucleotides of the RNA strand in RNA/DNA hybrids, and Pfu DNA polymerase can extend the 3'-mismatched RNA primer after the 3'-mismatched ribonucleotide is removed by PfRecJ. Finally, we reconstituted the primer-proofreading reaction of a 3'-mismatched ribonucleotide RNA/DNA hybrid using PfRecJ, replication protein A, Proliferating cell nuclear antigen (PCNA) and PolB. Given that PfRecJ is associated with the GINS complex, a central nexus in archaeal DNA replication fork, we speculate that PfRecJ proofreads the RNA primer in vivo.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Taq-DNA-Polymerase aus Thermus aquaticus, with 10× PCR reaction buffer containing MgCl2
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Taq-DNA-Polymerase aus Thermus aquaticus, with 10× PCR reaction buffer without MgCl2
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
DNA-Polymerase I, Klenow-Fragment aus E. coli, buffered aqueous glycerol solution
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
DNA-Polymerase I aus E. coli lysogen für NM 964, buffered aqueous glycerol solution
Preise und Verfügbarkeit sind derzeit nicht verfügbar.