Merck
  • Comparison of properties of tablets and energy profile of compaction of two spray-dried lactoses.
Comparison of properties of tablets and energy profile of compaction of two spray-dried lactoses.
Acta poloniae pharmaceutica (2013-04-25)
Jitka Muzíková, Pavla Sináglová
ZUSAMMENFASSUNG
The paper compared two spray-dried lactoses Flowlac 100 and SuperTab 14SD from the standpoint of tensile strength and disintegration time of tablets, the effect of an addition of the lubricant magnesium stearate and silicified microcrystalline cellulose on these properties, and also from the standpoint of the energy profile of compression. The comparison of the values was performed at the compression force of 15 kN. The strength of tablets was higher in the case of SuperTab 14SD, an increase in the concentration of magnesium stearate did not decrease tablet strength. Prosolv SMCC 90 increased the strength of tablets and made it equal for both lactoses, but it also increased the sensitivity to the added lubricant. The disintegration time of tablets was shorter in the case of SuperTab 14SD, an increased concentration of magnesium stearate prolonged it, and an addition of Prosolv SMCC 90 shortened it and made it equal for both lactoses. From the energy standpoint, the maximal energy was higher in the case of SuperTab 14SD, an addition of Prosolv SMCC 90 increased it and again made it equal for both lactoses. The differences in the values of the maximal energy were primarily due to the values of the energy for friction and the energy accumulated by the tablet after compression, and there was no marked difference in the values of the energy of decompression. SuperTab 14SD showed a higher plasticity than Flowlac 100.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cellulosepulver, microcrystalline, powder, 20 μm
Sigma-Aldrich
Stearinsäure, Grade I, ≥98.5% (capillary GC)
Sigma-Aldrich
Cellulosepulver, microcrystalline, powder
Sigma-Aldrich
Cellulosepulver, fibers, (medium)
Sigma-Aldrich
Stearinsäure, reagent grade, 95%
Sigma-Aldrich
Sigmacell Cellulose, Type 20, 20 μm
Supelco
Avicel® PH-101, ~50 μm particle size
Sigma-Aldrich
α-Cellulose, powder
Sigma-Aldrich
Stearinsäure, ≥95%, FCC, FG
Supelco
Lactose, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Lactose, tested according to Ph. Eur.
Supelco
Stearinsäure, analytical standard
Sigma-Aldrich
Cellulosepulver, colloidal, microcrystalline
Supelco
Stearinsäure, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sigmacell Cellulose, Type 50, 50 μm
Sigma-Aldrich
Sigmacell Cellulose, Type 101, Highly purified, fibers
Sigma-Aldrich
Avicel® PH-101, tested according to Ph. Eur.
Sigma-Aldrich
α-Cellulose, BioReagent, suitable for insect cell culture
Stearinsäure, European Pharmacopoeia (EP) Reference Standard
Supelco
Cellulosepulver, powder, suitable for column chromatography
Lactose, wasserfrei, European Pharmacopoeia (EP) Reference Standard
Supelco
Stearinsäure, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Cellulosepulver, DS-0, powder, suitable for thin layer chromatography (TLC)
Supelco
Cellulosepulver, acid washed, powder, suitable for column chromatography
Supelco
Cellulosepulver, DFS-0, microcrystalline, suitable for thin layer chromatography (TLC)
Supelco
Cellulosepulver, acid washed, from spruce, suitable for column chromatography