Merck
Faecalibacterium prausnitzii and Crohn's disease - is there any connection?
Polish journal of microbiology (2013-07-09)
Mirosława Gałecka, Patrycja Szachta, Anna Bartnicka, Liliana Łykowska-Szuber, Piotr Eder, Andreas Schwiertz
ZUSAMMENFASSUNG
The aim of the study was evaluation of the correlation between the level of clinical activity of Crohn's disease (CD) and the number of Faecalibacterium prausnitzii, total number of bacteria and the concentration of selected short chain fatty acids (SCFA) in stool. 34 individuals diagnosed with Crohn's disease participated in this study in 2011. The disease activity was determined according to the Crohn Disease Activity Index (CDAI). The number of Faecalibacterium prausnitzii and total number of bacteria were monitored by RT-PCR. The concentrations of SCFA were determined by gas chromatography. In CD patients, Faecalibacterium prausnitzii number and percentage of the total number of bacteria were greatly reduced. In patients with CD the percentage of acetate was elevated (70%), while the percentages of propionate and butyrate were significantly reduced (14.9% and 7.99%, respectively).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Buttersäure, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Buttersäure, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Buttersäure, ≥99%, FG
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Buttersäure, natural, ≥99%, FCC, FG
Preise und Verfügbarkeit sind derzeit nicht verfügbar.