Merck
  • Calcium phosphate/octadecyl-quatemized carboxymethyl chitosan nanoparticles: an efficient and promising carrier for gene transfection.
Calcium phosphate/octadecyl-quatemized carboxymethyl chitosan nanoparticles: an efficient and promising carrier for gene transfection.
Journal of nanoscience and nanotechnology (2013-07-26)
Ying Sun, Xiaoyu Li, Xiaofei Liang, Zhiyong Wan, Yourong Duan
ZUSAMMENFASSUNG
Calcium phosphate (CaP) has been widely used as the vector for gene transfection in the past three decades. However, clinical application is still not popular due to the poor-controlling of DNA/CaP complexes preparation, cytotoxicity and its low transfection efficiency. In this study, a novel amphipathic octadecyl-quatemized carboxymethyl chitosan (OQCMC) derivative from chitosan was combined with calcium phosphate to synthesize CaP/OQCMC nanoparticles (CaP/OQCMC NPs). The nanoparticles were 122-177 nm in diameter exhibited neutral zeta potential (from -0.115 mV to 0.216 mV), and they were applied as DNA vectors for DNA loading and in vitro transfection. The results showed that CaP/OQCMC NPs displayed high DNA loading capacity and enhanced transfection efficiency with extremely low cytotoxicity. In addition, both CaP and OQCMC are biocompatible and biodegradable, thus the as-prepared CaP/OQCMC NPs are promising in gene delivery.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Chitosan, low molecular weight
Sigma-Aldrich
Chitosan, medium molecular weight
Sigma-Aldrich
Chitosan, high molecular weight
Sigma-Aldrich
Chitosan, from shrimp shells, ≥75% (deacetylated)
Sigma-Aldrich
Chitosan, from shrimp shells, practical grade
Sigma-Aldrich
Desoxyribonukleinsäure, einsträngig aus Lachshoden, For hybridization
Sigma-Aldrich
Desoxyribonukleinsäure, einsträngig aus Lachshoden, For hybridization
Sigma-Aldrich
Chitosan aus Garnelenschalen, low-viscous
Sigma-Aldrich
Desoxyribonukleinsäure Natriumsalz aus Heringshoden, Type XIV
Sigma-Aldrich
Plasmid DNA from E. coli RRI, pUC19, buffered aqueous solution
Sigma-Aldrich
Plasmid DNA from E. coli RRI, pUC18, buffered aqueous solution
Sigma-Aldrich
Desoxyribonukleinsäure aus menschlicher Placenta, buffered aqueous solution, sexed, female