Merck
Effect of water soluble carrier on dissolution profiles of diclofenac sodium.
Acta poloniae pharmaceutica (2013-08-08)
Barbara Cwiertnia
ZUSAMMENFASSUNG
Pharmaceutical aviailability of diclofenac sodium from solid dispersions of PEG 6000 have been studied in comparison to those of the corresponding physical mixtures and pure diclofenac sodium. The diclofenac sodium is poorly water soluble drug. The properties of diclofenac sodium-PEG 6000 solid dispersions have been determined by the methods of differential scanning calorimetry (DSC), X-ray diffraction and scanning electron microscopy (SEM). The effect of PEG 6000 on the solubility of selected diclofenac sodium dispersions has been studied. The solubility of diclofenac sodium from its solid dispersion has been found to increase in the presence of PEG 6000.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasser, suitable for HPLC
Sigma-Aldrich
Wasser, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Wasser, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Wasser, Deionized
Supelco
Wasser, suitable for ion chromatography
Sigma-Aldrich
Wasser, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Wasser, Molecular Biology, sterile filtered
Supelco
Wasser, for TOC analysis
Sigma-Aldrich
Diclofenac Natriumsalz
Sigma-Aldrich
Wasser, BioPerformance Certified
Supelco
Diclofenac Natriumsalz, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Wasser, ACS reagent
Supelco
Wasser, ACS reagent, for ultratrace analysis
Sigma-Aldrich
Wasser, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Wasser, PCR Reagent
Sigma-Aldrich
Wasser, tested according to Ph. Eur.
Supelco
Wasser, suitable for HPCE, suitable for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Wasser, endotoxin, free
Diclofenac-Natrium, European Pharmacopoeia (EP) Reference Standard
Supelco
Dichte Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Diclofenac-Kalium, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Wasser-16O, ≥99.94 atom % 16O