Merck
  • Removal of Pb(II) from aqueous solution by hydrous manganese dioxide: adsorption behavior and mechanism.
Removal of Pb(II) from aqueous solution by hydrous manganese dioxide: adsorption behavior and mechanism.
Journal of environmental sciences (China) (2013-08-08)
Meng Xu, Hongjie Wang, Lei Di, Dan Qu, Yujia Zhai, Yili Wang
ZUSAMMENFASSUNG
Hydrous manganese dioxide (HMO) synthesized by redox of potassium permanganate and hydrogen peroxide was used as an adsorbent for Pb(II) removal. The specific surface area, pore volume and BJH pore diameter of the HMO were 79.31 m2/g, 0.07 cm3/g and 3.38 nm, respectively. The adsorption equilibrium at 298 K could be well described by the Langmuir isotherm equation with qmax value of 352.55 mg/g. The negative values of deltaG and the positive values of deltaH and deltaS indicated the adsorption process was spontaneous and endothermic. The pseudo second-order equation could best fit the adsorption data. The value of the calculated activation energy for Pb(II) adsorption onto the HMO was 38.23 kJ/mol. The uptake of Pb(II) by HMO was correlated with increasing surface hydroxyl group content and the main adsorbed speciation was PbOH+. The final chemical state of Pb(II) on the surface of HMO was similar to PbO. HMO was a promising candidate for Pb(II) removal from aqueous solution.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasser, suitable for HPLC
Sigma-Aldrich
Wasser, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Wasser, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Wasser, Deionized
Supelco
Wasser, suitable for ion chromatography
Sigma-Aldrich
Mangan(IV)-oxid, ReagentPlus®, ≥99%
Sigma-Aldrich
Wasser, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Wasser, Molecular Biology, sterile filtered
Sigma-Aldrich
Mangan(IV)-oxid, activated, ~85%, <10 μm
Supelco
Wasser, for TOC analysis
Sigma-Aldrich
Wasser, BioPerformance Certified
Sigma-Aldrich
Wasser, ACS reagent
Sigma-Aldrich
Mangan(IV)-oxid, 10 μm, reagent grade, ≥90%
Sigma-Aldrich
Stickstoff, ≥99.998%
Sigma-Aldrich
Blei, powder, −100 mesh, 99.95% trace metals basis
Supelco
Wasser, ACS reagent, for ultratrace analysis
Sigma-Aldrich
Wasser, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Wasser, PCR Reagent
Sigma-Aldrich
Wasser, tested according to Ph. Eur.
Supelco
Wasser, suitable for HPCE, suitable for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Blei, shot, <2 mm, 99.9% trace metals basis
Sigma-Aldrich
Wasser, endotoxin, free
Sigma-Aldrich
Blei, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Blei, shot, 1-3 mm, 99.995% trace metals basis
Supelco
Dichte Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Wasser-16O, ≥99.94 atom % 16O