Merck
  • [Changes in the content of plasma lipoproteins in persons subjected to diets prepared with sunflower oil alone or mixed with palm olein].
[Changes in the content of plasma lipoproteins in persons subjected to diets prepared with sunflower oil alone or mixed with palm olein].
Investigacion clinica (2013-08-21)
María Isabel Giacopini de Z, Hilda V Alonso, Josefina Sánchez, Ninoska García, Lilia Veliz, Iván Golfetto, Virgilio Bosch
ZUSAMMENFASSUNG
We analyzed in 31 subjects, regular guests of the University food service of the Central University of Venezuela (UCVFS), in Caracas, the effects of replacing sunflower oil, commonly used in the preparation of meals, by a mix of sunflower oil and palm olein 70/30 (v/v) respectively. Plasma concentrations of total cholesterol, low and very low density lipoproteins were not changed after 40 days of the substitution. On the contrary, concentrations of high density lipoprotein and total triglycerides increased. The resistance to the oxidation of low-density lipoproteins increased considerably (p < 0.01). Today this resistance is considered as a protective factor of great importance in the prevention of the initiation of the atherogenic process. Taking into account the favorable modifications of HDL cholesterol and the clear increased resistance to the oxidation of LDL, we think that palm olein, mixed with other oils with a high ratio linoleic/palmitic (sunflower, corn, soya an the likes), can be used as a healthy alternative in human nutrition.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Palmitinsäure, ≥99%
Sigma-Aldrich
α-Linoleic acid, ≥98%
Sigma-Aldrich
Sonnenblumenkernöl aus Helianthus annuus
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Laurinsäure, ≥98%, FCC, FG
Sigma-Aldrich
Linolsäure, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Palmitinsäure, BioXtra, ≥99%
Sigma-Aldrich
Myristinsäure, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Supelco
Palmitinsäure, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Laurinsäure, 98%
Sigma-Aldrich
Natrium-dodecanoat, 99-100%
Supelco
Cholesterin -Lösung, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Palmitinsäure, ≥98%, FCC, FG
Sigma-Aldrich
Linolsäure, technical, 58-74% (GC)
Supelco
Linolsäure, analytical standard
Sigma-Aldrich
Linolsäure Natriumsalz, ≥97% (GC)
Supelco
Palmitinsäure, analytical standard
Sigma-Aldrich
Laurinsäure, natural, ≥98%, FCC, FG
Sigma-Aldrich
Dodecansäure, ≥99% (GC/titration)
Supelco
Palmöl, analytical standard
Sigma-Aldrich
Myristinsäure, natural, ≥98.5%, FG
Supelco
Sonnenblumenöl, analytical standard
Sigma-Aldrich
Palmitinsäure, ≥98% palmitic acid basis (GC)
Sigma-Aldrich
Myristinsäure, ≥95%, FCC, FG
Sigma-Aldrich
Myristinsäure, ≥98.0% (GC)
Palmitinsäure, European Pharmacopoeia (EP) Reference Standard
Supelco
Myristinsäure, analytical standard