- Gas-liquid chromatographic determination of ethopabate in feed premixes.
Gas-liquid chromatographic determination of ethopabate in feed premixes.
Journal - Association of Official Analytical Chemists (1978-05-01)
R L Smallidge
PMID649547
ZUSAMMENFASSUNG
A gas-liquid chromatographic method is presented for determining ethopabate in 0.8 and 8.0% premixes. A sample is extracted with tetrahydrofuran containing an internal standard, by sonication or overnight soaking. The extract is clarified by centrifugation, diluted if necessary, and injected into a gas chromatograph equipped with a flame ionization detector. Average per cent recoveries for spiked blank samples were 100.6 at the 0.8% level and 100.4 at the 8.0% level. Precision, as indicated by replicate analyses of several premixes, ranged from 0.5 to 1.7% relative standard deviation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Preise und Verfügbarkeit sind derzeit nicht verfügbar.