Merck
[Effectiveness of non-starch polysaccharide crylan in experimental hyperlipidemia].
Voprosy pitaniia (1995-01-01)
O V Remezova, I V Okunevich, G A Vitovskaia, E P Anan'eva, V E Ryzhenkov
ZUSAMMENFASSUNG
In vitro experiments were shown that native ecologically pure non-starch polysaccharide crylan bind the bile acid. Crylan was found to decrease hyperlipidemia induced in rats by means of diet enriched with cholesterol and 6-methylthiouracil. This effect of crylan was more pronounced as compare with the action of native nonspecific enterosorbent polyfepan.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
6-Methyl-2-thiouracil, VETRANAL®, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
6-Methyl-2-thiouracil, purum, ≥98.0% S basis (elemental analysis)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.