Merck
  • Extrathyroidally mediated changes in circulating thyroid hormone concentrations in the male rat following administration of an experimental oxyacetamide (FOE 5043).
Extrathyroidally mediated changes in circulating thyroid hormone concentrations in the male rat following administration of an experimental oxyacetamide (FOE 5043).
Toxicology and applied pharmacology (1995-06-01)
W R Christenson, B D Becker, H D Hoang, B S Wahle, K D Moore, P D Dass, S G Lake, B P Stuart, D L Van Goethem, G K Sangha
ZUSAMMENFASSUNG
Evidence of increased hepatic metabolizing capacity coupled with reductions in serum thyroxine (T4) levels were noted in the rat during preliminary toxicity studies with FOE 5043, an oxyacetamide with herbicidal properties. These findings were consistent with reports in the literature suggesting that declines in T4 as a result of exposure to various classes of chemicals may be mediated extrathyroidally, such as through chemical induction of hepatic thyroid hormone metabolism. To examine this question with respect to FOE 5043, male rats were surgically thyroidectomized and provided thyroid hormone replacement therapy via implanted osmotic minipumps capable of maintaining a T4/triiodothyronine (T3) serum concentration for approximately 4 weeks at a level comparable to that of euthyroid controls. Seven days after minipump implantation, thyroidectomized + T4/T3 (TX + T4/T3) and nonthyroidectomized intact rats (NTX) were fed diets containing 0, 25, 1000, or 3000 ppm FOE 5043 for up to 3 weeks. Dose-related and equivalent declines in total and free serum T4 levels in both TX + T4/T3 and NTX rats were measured at Weeks 1, 2, and 3. Alterations in thyrotropin, total, free, and reverse serum T3 levels were also noted in both TX and NTX animals; however, a compound-related trend was difficult to discern and, when compared to the T4 response, the changes were markedly less consistent with respect to both time and dose. Additionally, dose-related increases in absolute and relative liver weights were measured in both TX + T4/T3 and NTX animals. As the only source of thyroid hormone in the TX + T4/T3 animals was that provided by the pump, these data suggest that FOE 5043-induced alterations in serum thyroid hormone levels, most notably T4, are being mediated indirectly, possibly as a result of increased hepatic metabolism, rather than through a direct effect on the thyroid gland.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Flufenacet, PESTANAL®, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.