Merck
  • Matrix-assisted laser desorption/ionization mass spectrometry of restriction enzyme-digested plasmid DNA using an active Nafion substrate.
Matrix-assisted laser desorption/ionization mass spectrometry of restriction enzyme-digested plasmid DNA using an active Nafion substrate.
Rapid communications in mass spectrometry : RCM (1994-09-01)
J Bai, Y H Liu, D M Lubman, D Siemieniak
ZUSAMMENFASSUNG
Matrix-assisted laser desorption/ionization using a 3-hydroxypicolinic acid matrix from an active Nafion substrate has been used for detection of restriction enzyme-digested double-stranded plasmid DNA using time-of-flight mass spectrometry. DNA strands of up to 267 base pairs were detected with minimal sample purification, although only as species corresponding to single-stranded DNA.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3-Hydroxypyridin-2-carboxylsäure, 98%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
3-Hydroxypicolsäure, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.