Merck
  • Matrix-degrading lysosomal hydrolases in serum of rats with hepatic fibrosis treated with methylthiouracil or thyroideum.
Matrix-degrading lysosomal hydrolases in serum of rats with hepatic fibrosis treated with methylthiouracil or thyroideum.
Romanian journal of endocrinology (1993-01-01)
K Olczyk, E J Kucharz, P Sonecki
ZUSAMMENFASSUNG
Thyroid hormones or methylthiouracil (MTU) were given to healthy rats or animals with hepatic fibrosis. Administration of thyroid hormones to rats with carbon tetrachloride-induced hepatic fibrosis caused an increase in serum activity of matrix-degrading hydrolases although the increase was lower than that in healthy animals treated with thyroid hormones only. Methylthiouracil administration had no effect on the enzyme activity in healthy rats and resulted in a decreased activity in rats with hepatic fibrosis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
6-Methyl-2-thiouracil, VETRANAL®, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
6-Methyl-2-thiouracil, purum, ≥98.0% S basis (elemental analysis)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.