Merck
  • Overall migration and specific migration of bisphenol A diglycidyl ether monomer and m-xylylenediamine hardener from an optimized epoxy-amine formulation into water-based food simulants.
Overall migration and specific migration of bisphenol A diglycidyl ether monomer and m-xylylenediamine hardener from an optimized epoxy-amine formulation into water-based food simulants.
Food additives and contaminants (1993-09-01)
J Simal Gándara, P López Mahía, P Paseiro Losada, J Simal Lozano, S Paz Abuín
ZUSAMMENFASSUNG
The overall and specific migrations of BADGE n = 0 monomer and m-XDA hardener from a BEPOX LAB 889 (Gairesa internal code), epoxy system cured at room temperature, into three water-based food simulants are studied. Hydrolysis of BADGE n = 0 was observed in all of these simulants, giving more polar products. We thus propose changing the EEC Directives, which at present only legislate for levels of BADGE n = 0 monomer in the simulants, to include the hydrolysis products of BADGE monomers. Another alternative would be to express all the migration levels due to BADGE and its derived products in terms of BADGE itself.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
m-Xylylenediamin, 99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.