Merck
  • Effects of zaldaride maleate, an antidiarrheal compound, on intracellular Ca2+-mediated short-circuit current response in the rat colonic mucosa.
Effects of zaldaride maleate, an antidiarrheal compound, on intracellular Ca2+-mediated short-circuit current response in the rat colonic mucosa.
Japanese journal of pharmacology (2000-07-11)
N Aikawa, A Karasawa, K Ohmori
ZUSAMMENFASSUNG
The effect of zaldaride maleate (ZAL) on intracellular Ca2+-mediated short-circuit current (Isc) response in rat colonic mucosa was investigated. The Isc response to acetylcholine was partially reduced by the neuronal blocker tetrodotoxin. The tetrodotoxin-sensitive Isc response to acetylcholine was inhibited by ZAL. The tetrodotoxin-insensitive effect, which indicates direct stimulation of acetylcholine in colonic mucosa, was not affected by ZAL. The Isc response to the Ca2+ ionophore A23187 was reduced by ZAL in the presence of tetrodotoxin. These results suggest that ZAL reduces colonic ion secretion by inhibiting a tetrodotoxin-sensitive neuronal pathway and an intracellular Ca2+ pathway in epithelium.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
CGS-9343B, ≥98% (HPLC), powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.