Merck
Intestinal metabolism of mephentermine and its biliary metabolites in male Wistar rats.
Xenobiotica; the fate of foreign compounds in biological systems (1992-06-01)
M Mori, M Kobayashi, H Uemura, T Miyahara, H Kozuka
ZUSAMMENFASSUNG
1. Intestinal metabolites produced in the incubation (0-24 h) of mephentermine (MP), phentermine (Ph), N-hydroxymephentermine (N-hydroxy-MP), N-hydroxyphentermine (N-hydroxy-Ph), p-hydroxymephentermine (p-hydroxy-MP) and p-hydroxyphentermine (p-hydroxy-Ph) with male Wistar rat intestinal contents under N2 were examined by g.l.c. and g.l.c.-electron impact (EI) mass spectrometry. Metabolites produced in the anaerobic incubation of bile from rats given MP, with the intestinal contents were also examined. In addition, urinary and biliary metabolites of p-hydroxy-MP and p-hydroxy-Ph dosed orally to rat were examined. 2. Metabolites in the anaerobic incubation of N-hydroxy-MP and N-hydroxy-Ph were MP and Ph, and Ph, respectively. No metabolites were detected in the incubation of MP, Ph, p-hydroxy-MP and p-hydroxy-Ph. 3. p-Hydroxy-MP and p-hydroxy-Ph (major), and MP and Ph (minor) were detected when bile from rats given MP was incubated with intestinal contents. 4. Unchanged p-hydroxy-MP, and conjugates of p-hydroxy-MP and p-hydroxy-Ph, were detected in the 24-h urine of rats dosed with p-hydroxy-MP, which accounted for about 3, 72 and 1% dose, respectively. Unchanged p-hydroxy-Ph and conjugated p-hydroxy-Ph were detected in the 24-h urine of rats dosed with p-hydroxy-Ph, which accounted for about 4 and 68% dose, respectively. 5. Conjugated p-hydroxy-MP and conjugated p-hydroxy-Ph, which accounted for about 3% doses, were detected in the 24-h bile of rats dosed with p-hydroxy-MP and p-hydroxy-Ph.(ABSTRACT TRUNCATED AT 250 WORDS)