Merck
The role of histone acetylation in SMN gene expression.
Human molecular genetics (2005-03-18)
Lauren E Kernochan, Melissa L Russo, Nathaniel S Woodling, Thanh N Huynh, Amy M Avila, Kenneth H Fischbeck, Charlotte J Sumner
ZUSAMMENFASSUNG
Increasing survival motor neuron 2 (SMN2) gene expression may be an effective strategy for the treatment of spinal muscular atrophy (SMA). Histone deacetylase (HDAC) inhibitors have been shown to increase SMN transcript and protein levels, but the specific role of histone acetylation in regulating SMN gene expression has not been explored. Using chromatin immunopreciptation, we investigated the levels of acetylated H3 and H4 histones and HDACs associated with different regions of the human and mouse SMN genes in both cultured cells and tissues. We show that the SMN gene has a reproducible pattern of histone acetylation that is largely conserved among different tissues and species. A limited region of the promoter surrounding the transcriptional start site has relatively high levels of histone acetylation, whereas regions further upstream or downstream have lower levels. After HDAC inhibitor treatment, acetylated histone levels increased, particularly at upstream regions, correlating with a 2-fold increase in promoter activity. During development in mouse tissues, histone acetylation levels decreased and associated HDAC2 levels increased at the region closest to the transcriptional start site, correlating with a 40-60% decrease in SMN transcript and protein levels. These data indicate that histone acetylation modulates SMN gene expression and that pharmacological manipulation of this epigenetic determinant is feasible. HDAC2, in particular, may be a future therapeutic target for SMA.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Acetyl-Histon-H3-Antikörper, from rabbit
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Valpromid, ≥97% (NMR)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.