- Cholecystocolonic fistula diagnosed with CT-intravenous cholangiography.
Cholecystocolonic fistula diagnosed with CT-intravenous cholangiography.
Australasian radiology (2006-09-20)
F Gaillard, D Stella, R Gibson
PMID16981948
ZUSAMMENFASSUNG
Cholecystoenteric fistulas are often not diagnosed preoperatively and delineation of fistula can have an influence on surgical planning. We report a case of cholecystocolonic fistula diagnosed preoperatively using CT-i.v. cholangiography and review the published reports.