Merck
The mechanism and kinetics of decomposition of 5-aminotetrazole.
Journal of molecular modeling (2008-03-12)
Jian-Guo Zhang, Li-Na Feng, Shao-Wen Zhang, Tong-Lai Zhang, Hui-Hui Zheng
ZUSAMMENFASSUNG
The pathway and ab initio direct kinetics of the decomposition 5-aminotetrazole (5-ATZ) to HN(3) and NH(2)CN was investigated. Reactant, products and transition state were optimized with MP2 and B3LYP methods using 6-311G** and aug-cc-pVDZ basis sets. The intrinsic reaction coordinate curve of the reaction was calculated using the MP2 method with 6-311G** basis set. The energies were refined using CCSD(T)/6-311G**. Rate constants were evaluated by conventional transition-state theory (CVT) and canonical variational transition-state theory (TST), with tunneling effect over 300 to 2,500 K. The results indicated that the tunneling effect and the variational effect are small for the calculated rate constants. The fitted three-parameter expression calculated using the CVT and TST methods are k(T) = 4.07 x 10(11) x T(0.84) x e((-2.42 x 10(4))/T)s(-1) and k(T) = 2.09 x 10(11) x T(0.89) x e((-2.36 x 10(4))/T)s(-1), respectively.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
5-Aminotetrazol, 97%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.