Merck
  • Electrochemical genotyping and detection of single-nucleotide polymorphisms based on junction-probe containing 2'-deoxyinosine.
Electrochemical genotyping and detection of single-nucleotide polymorphisms based on junction-probe containing 2'-deoxyinosine.
Chemical communications (Cambridge, England) (2010-08-24)
Jing Zhang, Xiaoyan Wu, Pingping Chen, Ningting Lin, Jinghua Chen, Guonan Chen, Fengfu Fu
ZUSAMMENFASSUNG
A novel electrochemical biosensor for the genotyping/detection of single-nucleotide polymorphisms (SNPs) of trace oral-cancer salivary DNA based on junction probes containing 2'-deoxyinosine (dI) residues is described.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2′-Desoxyinosin, ≥98%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.