Merck
Effects of tacrolimus ointment on Toll-like receptors in atopic dermatitis.
Clinical and experimental dermatology (2010-11-13)
E Antiga, W Volpi, D Torchia, P Fabbri, M Caproni
ZUSAMMENFASSUNG
Alterations of Toll-like receptors (TLRs) seem to play a role in susceptibility to atopic dermatitis (AD). To investigate the expression of TLRs in moderate to severe chronic AD in adults before and after a 3-week treatment with 0.1% tacrolimus ointment, compared with 0.1% topical hydrocortisone-17-butyrate. In total, 21 adult patients with AD were enrolled: 11 were given tacrolimus ointment and 10 were given hydrocortisone butyrate; a further 6 healthy adults formed the control group. The clinical efficacy of the treatment was assessed using the SCORing Atopic Dermatis (SCORAD) index. Biopsies were taken from lesional skin before and after treatment, which were stained immunohistochemically with monoclonal antibodies to TLR-1, -2, -4 and -9. Both 3-week topical treatments improved signs and symptoms in all 21 patients considered, with no significant difference between the two groups. In the skin of patients with AD, TLR-1 was overexpressed and TLR-2 underexpressed compared with healthy controls, whereas no differences were found for TLR-4 and TLR-9. Staining for TLR-1 was decreased in both groups after treatment. AD specimens had higher levels of TLR-2 expression after either treatment compared with baseline, and levels were higher after tacrolimus treatment than after hydrocortisone butyrate. Neither tacrolimus nor hydrocortisone butyrate affected expression of TLR-4 or TLR-9. Short-term therapy with tacrolimus ointment reduced expression of TLR-1, which may inhibit the antimicrobial potential of TLR-2, and also reversed the impairment of TLR-2 in AD lesions. Expression of TLR-4 and TLR-9 was not affected by tacrolimus.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Hydrocortisone 17-butyrate
Preise und Verfügbarkeit sind derzeit nicht verfügbar.