Merck
  • Stampidine as a promising antiretroviral drug candidate for pre-exposure prophylaxis against sexually transmitted HIV/AIDS.
Stampidine as a promising antiretroviral drug candidate for pre-exposure prophylaxis against sexually transmitted HIV/AIDS.
Expert opinion on investigational drugs (2012-03-01)
Fatih M Uckun, Pedro Cahn, Sanjive Qazi, Osmond D'Cruz
ZUSAMMENFASSUNG
Pre-exposure prophylaxis (PrEP) is an evolving new approach to prevention of sexually transmitted HIV-1 that employs antiretroviral (ARV) agents prior to potential HIV-1 exposure in an attempt to reduce the likelihood of HIV-1 infection postexposure. The identification of new ARV agents with potent activity against multidrug-resistant HIV remains an unmet and urgent challenge in the field of PrEP. This article reviews the preclinical and early clinical activity and safety profile of stampidine, a novel antiretroviral (ARV) drug candidate that exhibits remarkable subnanomolar to low nanomolar in vitro antiretroviral potency against genotypically and phenotypically nucleoside reverse transcriptase inhibitor (NRTI)-resistant primary clinical HIV isolates, non-nucleoside RT-resistant HIV-1 isolates. Stampidine has a favorable pharmacokinetic profile in mice, rats, dogs and cats with 25 or 50 mg/kg tolerable dose levels yielding micromolar plasma concentrations that are 1000-fold higher than its in vitro IC(50) value against HIV. Stampidine has a favorable, safety profile in mice, rats, dogs and cats and it showed significant in vivo ARV activity in HIV-infected Hu-PBL-SCID mice as well as FIV-infected domestic cats. Furthermore, it did not cause any maternal toxicity, developmental toxicity or teratogenicity in rabbits treated at 10 - 40 mg/kg/day dose levels. In a recently completed first-in-human Phase I clinical trial, stampidine did not cause dose-limiting toxicity at single dose levels ranging from 5 to 25 mg/kg. The favorable safety and activity profile of stampidine warrants its further development as a promising next-generation PrEP candidate to prevent the sexual transmission of HIV-1. The discovery of stampidine as a potent antiretroviral agent represents a significant step forward in the development of effective therapeutic as well as preventive strategies against HIV/AIDS.