Merck
Design, synthesis and molecular docking studies of sinomenine derivatives.
Bioorganic & medicinal chemistry letters (2012-08-16)
Xiaoyun Chai, Zhongjun Guan, Shichong Yu, Qingjie Zhao, Honggang Hu, Yan Zou, Xia Tao, Qiuye Wu
ZUSAMMENFASSUNG
In order to search for drugs with excellent anti-inflammatory activities, a series of novel sinomenine derivatives were designed, synthesized, and evaluated for their inhibition activities against NF-κB activation induced by lipopolysaccharide (LPS). Compared with the natural parent sinomenine, compounds 2a-w showed higher activity, while compounds 1a-o showed similar activity against NF-κB. Moreover, a molecular model for the binding between compound 2v and the active site of p50 was provided on the basis of the computational docking results.